Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGSF1
Entrez Gene ID
3547
Gene Name
immunoglobulin superfamily member 1
Gene Aliases
CHTE, IGCD1, IGDC1, INHBP, PGSF2, p120
Chromosome Location
Chr.X: 131273506 - 131289457 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170963.1 NP_001164434.1 1 - 2 42 77

View Full Product Details