The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171135.1 | NP_001164606.1 | 1 - 2 | 236 | 55 |
GenBank mRNA | BC015030.1 | - | 1 - 2 | 236 | 55 |