Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKX3-2
Entrez Gene ID
579
Gene Name
NK3 homeobox 2
Gene Aliases
BAPX1, NKX3.2, NKX3B, SMMD
Chromosome Location
Chr.4: 13540830 - 13544490 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001189.3 NP_001180.1 1 - 2 546 90
GenBank mRNA AF009801.1 - 1 - 2 470 90
BC111966.1 - 1 - 2 546 90

View Full Product Details