Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT85
Entrez Gene ID
3891
Gene Name
keratin 85
Gene Aliases
ECTD4, HB5, Hb-5, K85, KRTHB5, hHb5
Chromosome Location
Chr.12: 52360006 - 52367525 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300810.1 NP_001287739.1 6 - 7 901 66
NM_002283.3 NP_002274.1 8 - 9 1455 66
GenBank mRNA AK302303.1 - 6 - 7 66
X99140.1 - 8 - 9 1401 66

View Full Product Details