The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294145.1 | NP_001281074.1 | 11 - 12 | 1635 | 101 |
NM_003142.4 | NP_003133.1 | 11 - 12 | 1311 | 101 | |
GenBank mRNA | AB451228.1 | - | 10 - 11 | 1137 | 101 |
AB451351.1 | - | 10 - 11 | 1137 | 101 | |
BC001289.1 | - | 11 - 12 | 1226 | 101 | |
BC020818.1 | - | 11 - 12 | 1244 | 101 | |
BT009862.1 | - | 10 - 11 | 1137 | 101 | |
J04205.1 | - | 11 - 12 | 1227 | 101 | |
M20328.1 | - | 8 - 9 | 978 | 101 | |
X13697.1 | - | 11 - 12 | 1209 | 101 | |
X69804.1 | - | 11 - 12 | 1385 | 101 |