The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003347.3 | NP_003338.1 | 1 - 2 | 225 | 76 |
NR_028436.2 | 1 - 2 | 225 | 76 | ||
XM_017028935.1 | XP_016884424.1 | 1 - 2 | 102 | 76 | |
GenBank mRNA | BC053368.1 | - | 1 - 2 | 78 | 76 |
BU584304.1 | - | 1 - 2 | 104 | 76 | |
CD690076.1 | - | 1 - 2 | 80 | 76 | |
CR456606.1 | - | 1 - 2 | 49 | 76 | |
DA271653.1 | - | 1 - 2 | 227 | 76 |