The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003824.3 | NP_003815.1 | 1 - 2 | 582 | 118 |
GenBank mRNA | AK291005.1 | - | 1 - 2 | 443 | 118 |
AY423721.1 | - | 1 - 2 | 118 | ||
BC000334.2 | - | 1 - 2 | 430 | 118 | |
BC025733.1 | - | 1 - 2 | 406 | 118 | |
BT006927.1 | - | 1 - 2 | 285 | 118 | |
CR456738.1 | - | 1 - 2 | 285 | 118 | |
U24231.1 | - | 1 - 2 | 414 | 118 | |
X84709.1 | - | 1 - 2 | 429 | 118 |