The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289161.1 | NP_001276090.1 | 1 - 2 | 212 | 68 |
NM_005931.4 | NP_005922.2 | 1 - 2 | 212 | 68 | |
GenBank mRNA | AK291927.1 | - | 1 - 2 | 210 | 68 |
AK314228.1 | - | 1 - 2 | 117 | 68 | |
BC044218.1 | - | 1 - 2 | 176 | 68 | |
DC427936.1 | - | 1 - 2 | 184 | 68 | |
X91625.1 | - | 1 - 2 | 73 | 68 |