Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT38
Entrez Gene ID
8687
Gene Name
keratin 38
Gene Aliases
HA8, KRTHA8, hHa8
Chromosome Location
Chr.17: 41436369 - 41441344 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006771.3 NP_006762.3 6 - 7 1664 81
GenBank mRNA AJ786656.1 - 6 - 7 1192 81
BC131715.1 - 6 - 7 1521 81

View Full Product Details