The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012318.2 | NP_036450.1 | 2 - 3 | 445 | 72 |
XM_006713884.1 | XP_006713947.1 | 2 - 3 | 153 | 72 | |
GenBank mRNA | AF061025.1 | - | 2 - 3 | 445 | 72 |
BC014500.1 | - | 2 - 3 | 345 | 72 | |
BC021208.1 | - | 2 - 3 | 370 | 72 |