The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014827.5 | NP_055642.3 | 3 - 4 | 501 | 79 |
XM_005245643.2 | XP_005245700.1 | 3 - 4 | 520 | 79 | |
XM_005245644.3 | XP_005245701.1 | 2 - 3 | 204 | 79 | |
XM_006711673.2 | XP_006711736.1 | 3 - 4 | 367 | 79 | |
XM_017002963.1 | XP_016858452.1 | 3 - 4 | 384 | 79 | |
GenBank mRNA | BC014268.2 | - | 3 - 4 | 339 | 79 |