The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015417.4 | NP_056232.2 | 4 - 5 | 586 | 99 |
GenBank mRNA | AL040853.1 | - | 2 - 3 | 200 | 99 |
BI520334.1 | - | 4 - 5 | 585 | 99 | |
EF560745.1 | - | 4 - 5 | 584 | 99 |