The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135037.1 | NP_001128509.1 | 1 - 2 | 51 | 89 |
GenBank mRNA | AK296468.1 | - | 1 - 2 | 51 | 89 |