The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143805.1 | NP_001137277.1 | 1 - 2 | 136 | 100 |
GenBank mRNA | AY054393.2 | - | 1 - 2 | 80 | 100 |
EF674517.1 | - | 1 - 2 | 136 | 100 |