Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BDNF
Entrez Gene ID
627
Gene Name
brain derived neurotrophic factor
Gene Aliases
ANON2, BULN2
Chromosome Location
Chr.11: 27654893 - 27722058 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143807.1 NP_001137279.1 1 - 2 235 101
GenBank mRNA AY054396.2 - 1 - 2 134 101
EF674520.1 - 1 - 2 235 101

View Full Product Details