The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143919.2 | NP_001137391.1 | 1 - 2 | 305 | 79 |
GenBank mRNA | D89079.1 | - | 1 - 2 | 305 | 79 |
U41070.1 | - | 1 - 2 | 288 | 79 |