The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143987.1 | NP_001137459.1 | 7 - 8 | 1058 | 92 |
GenBank mRNA | BC125161.1 | - | 7 - 8 | 1019 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011540763.1 | XP_011539065.1 | 7 - 8 | 1044 | 92 |