The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145207.1 | NP_001138679.1 | 3 - 4 | 448 | 78 |
NM_006620.3 | NP_006611.1 | 3 - 4 | 448 | 78 | |
GenBank mRNA | AF161394.1 | - | 3 - 4 | 373 | 78 |
AJ420551.1 | - | 3 - 4 | 365 | 78 | |
AJ459826.1 | - | 3 - 4 | 434 | 78 | |
AJ459827.1 | - | 3 - 4 | 434 | 78 | |
AK024258.1 | - | 3 - 4 | 369 | 78 | |
BC001465.2 | - | 3 - 4 | 357 | 78 | |
BC040849.1 | - | 3 - 4 | 352 | 78 | |
U87791.1 | - | 3 - 4 | 435 | 78 |