The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145207.1 | NP_001138679.1 | 4 - 5 | 634 | 108 |
XM_017010192.1 | XP_016865681.1 | 3 - 4 | 436 | 108 | |
GenBank mRNA | AF161394.1 | - | 4 - 5 | 559 | 108 |
AJ459827.1 | - | 4 - 5 | 620 | 108 |