The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146316.1 | NP_001139788.1 | 1 - 2 | 69 | 78 |
GenBank mRNA | BC071933.1 | - | 1 - 2 | 69 | 78 |