Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD34C
Entrez Gene ID
390616
Gene Name
ankyrin repeat domain 34C
Gene Aliases
-
Chromosome Location
Chr.15: 79282804 - 79298239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146341.1 NP_001139813.1 1 - 2 426 56
GenBank mRNA AK127037.1 - 1 - 2 426 56

View Full Product Details