The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016518.2 | NP_057602.2 | 1 - 2 | 443 | 80 |
GenBank mRNA | AF136970.1 | - | 1 - 2 | 129 | 80 |
AK022773.1 | - | 1 - 2 | 208 | 80 | |
AK027498.1 | - | 1 - 2 | 167 | 80 | |
BC008960.2 | - | 1 - 2 | 443 | 80 | |
BC027622.2 | - | 1 - 2 | 179 | 80 | |
CR457155.1 | - | 1 - 2 | 117 | 80 |