Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIPOX
Entrez Gene ID
51268
Gene Name
pipecolic acid and sarcosine oxidase
Gene Aliases
LPIPOX
Chromosome Location
Chr.17: 29042900 - 29057227 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016518.2 NP_057602.2 7 - 8 1367 84
XM_017024722.1 XP_016880211.1 6 - 7 966 84
GenBank mRNA AF134593.1 - 7 - 8 1084 84
AF136970.1 - 7 - 8 1053 84
AK022773.1 - 7 - 8 1132 84
AK027498.1 - 7 - 8 1091 84
BC008960.2 - 7 - 8 1367 84
BC027622.2 - 7 - 8 1103 84
CR457155.1 - 7 - 8 1041 84

View Full Product Details