The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018666.2 | NP_061136.2 | 18 - 19 | 2681 | 199 |
XM_017029621.1 | XP_016885110.1 | 18 - 19 | 2689 | 199 | |
XM_017029622.1 | XP_016885111.1 | 18 - 19 | 2600 | 199 | |
XM_017029623.1 | XP_016885112.1 | 18 - 19 | 2563 | 199 | |
XM_017029624.1 | XP_016885113.1 | 17 - 18 | 2503 | 199 | |
XM_017029625.1 | XP_016885114.1 | 17 - 18 | 2455 | 199 | |
XM_017029626.1 | XP_016885115.1 | 12 - 13 | 1805 | 199 | |
XM_017029627.1 | XP_016885116.1 | 8 - 9 | 1596 | 199 | |
GenBank mRNA | AJ278111.1 | - | 18 - 19 | 2681 | 199 |
BC144260.1 | - | 11 - 12 | 1434 | 199 | |
BF056519.1 | - | 2 - 3 | 199 |