The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018932.3 | NP_061755.1 | 1 - 1 | 155 | 82 |
GenBank mRNA | AK293336.1 | - | 1 - 1 | 177 | 82 |
AK316160.1 | - | 1 - 1 | 155 | 82 | |
BC045637.1 | - | 1 - 1 | 155 | 82 |