The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020382.4 | NP_065115.3 | 1 - 2 | 97 | 128 |
NR_136910.1 | 1 - 2 | 97 | 128 | ||
GenBank mRNA | AK292645.1 | - | 1 - 2 | 65 | 128 |
BC050346.1 | - | 1 - 2 | 440 | 128 |