The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021195.4 | NP_067018.2 | 1 - 2 | 40 | 59 |
GenBank mRNA | AK075329.1 | - | 1 - 2 | 38 | 59 |
BX444798.2 | - | 1 - 2 | 40 | 59 |