The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022843.3 | NP_073754.2 | 1 - 2 | 497 | 97 |
XM_017020708.1 | XP_016876197.1 | 3 - 4 | 1933 | 97 | |
GenBank mRNA | AF169693.1 | - | 1 - 2 | 123 | 97 |
AK094680.1 | - | 1 - 2 | 135 | 97 | |
AK289924.1 | - | 1 - 2 | 498 | 97 | |
AK314607.1 | - | 1 - 2 | 120 | 97 | |
AL833830.1 | - | 4 - 5 | 2239 | 97 | |
BC027907.1 | - | 1 - 2 | 121 | 97 |