Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C7orf25
Entrez Gene ID
79020
Gene Name
chromosome 7 open reading frame 25
Gene Aliases
-
Chromosome Location
Chr.7: 42909273 - 42913161 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024054.2 NP_076959.2 1 - 2 179 62

View Full Product Details