The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024886.2 | NP_079162.1 | 1 - 2 | 157 | 87 |
GenBank mRNA | AK024342.1 | - | 1 - 2 | 157 | 87 |
BC126459.1 | - | 1 - 2 | 102 | 87 |