Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM6
Entrez Gene ID
117854
Gene Name
tripartite motif containing 6
Gene Aliases
RNF89
Chromosome Location
Chr.11: 5596101 - 5612958 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058166.4 NP_477514.1 1 - 2 76 117
GenBank mRNA AF220030.1 - 1 - 2 64 117
AK027664.1 - 1 - 2 68 117
CR749260.1 - 1 - 2 49 117
DB233046.1 - 1 - 2 76 117

View Full Product Details