Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPCR1
Entrez Gene ID
135656
Gene Name
diffuse panbronchiolitis critical region 1
Gene Aliases
PBLT
Chromosome Location
Chr.6: 30941000 - 30954221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080870.3 NP_543146.2 1 - 2 80 79

View Full Product Details