The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145260.2 | NP_660303.1 | 1 - 2 | 269 | 81 |
GenBank mRNA | AK074591.1 | - | 1 - 2 | 281 | 81 |
BC025712.1 | - | 1 - 2 | 269 | 81 |