Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPERT
Entrez Gene ID
220082
Gene Name
spermatid associated
Gene Aliases
CBY2, NURIT
Chromosome Location
Chr.13: 45701030 - 45714559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152719.2 NP_689932.1 1 - 2 166 109
XM_011534971.2 XP_011533273.1 1 - 2 1440 109
GenBank mRNA AK093129.1 - 1 - 2 159 109
HY007057.1 - 1 - 2 158 109
HY017873.1 - 1 - 2 168 109

View Full Product Details