The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000883.3 | NP_000874.2 | 12 - 13 | 1345 | 91 |
NM_001102605.1 | NP_001096075.1 | 11 - 12 | 1315 | 91 | |
NM_001142573.1 | NP_001136045.1 | 9 - 10 | 1115 | 91 | |
NM_001142574.1 | NP_001136046.1 | 9 - 10 | 1100 | 91 | |
NM_001142575.1 | NP_001136047.1 | 8 - 9 | 1040 | 91 | |
NM_001142576.1 | NP_001136048.1 | 11 - 12 | 1246 | 91 | |
NM_001304521.1 | NP_001291450.1 | 9 - 10 | 1143 | 91 | |
NM_183243.2 | NP_899066.1 | 10 - 11 | 1237 | 91 | |
XM_017012168.1 | XP_016867657.1 | 11 - 12 | 1315 | 91 | |
XM_017012169.1 | XP_016867658.1 | 10 - 11 | 1237 | 91 | |
XM_017012170.1 | XP_016867659.1 | 9 - 10 | 1144 | 91 | |
XM_017012171.1 | XP_016867660.1 | 11 - 12 | 1433 | 91 | |
XM_017012172.1 | XP_016867661.1 | 11 - 12 | 1433 | 91 | |
GenBank mRNA | AK054640.1 | - | 8 - 9 | 1012 | 91 |
AK054667.1 | - | 10 - 11 | 1237 | 91 | |
AK092452.1 | - | 11 - 12 | 1282 | 91 | |
AK122994.1 | - | 11 - 12 | 1314 | 91 | |
AK293413.1 | - | 9 - 10 | 1072 | 91 | |
AK297943.1 | - | 9 - 10 | 1087 | 91 | |
BC033622.1 | - | 10 - 11 | 1228 | 91 | |
CD014008.1 | - | 10 - 11 | 1029 | 91 | |
CD014010.1 | - | 11 - 12 | 1151 | 91 | |
CR933672.1 | - | - | 1281 | 91 | |
J05272.1 | - | 12 - 13 | 1609 | 91 |