Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MBD3L5
- Entrez Gene ID
-
284428
- Gene Name
- methyl-CpG binding domain protein 3 like 5
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 7030583 - 7033011 on Build GRCh38
- Gene Symbol
- MBD3L3
- Entrez Gene ID
-
653657
- Gene Name
- methyl-CpG binding domain protein 3 like 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 7056205 - 7058634 on Build GRCh38
- Gene Symbol
- MBD3L4
- Entrez Gene ID
-
653656
- Gene Name
- methyl-CpG binding domain protein 3 like 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 7037744 - 7040173 on Build GRCh38
View Full Product Details