Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IFNA13
- Entrez Gene ID
-
3447
- Gene Name
- interferon alpha 13
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 21367372 - 21368076 on Build GRCh38
- Gene Symbol
- IFNA1
- Entrez Gene ID
-
3439
- Gene Name
- interferon alpha 1
- Gene Aliases
-
IFL, IFN, IFN-ALPHA, IFN-alphaD, IFNA13, IFNA@
- Chromosome Location
-
Chr.9: 21440454 - 21441316 on Build GRCh38
- Gene Symbol
- IFNA21
- Entrez Gene ID
-
3452
- Gene Name
- interferon alpha 21
- Gene Aliases
-
IFN-alphaI, LeIF F, leIF-F
- Chromosome Location
-
Chr.9: 21165637 - 21166660 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
V00540.1 |
- |
1 - 1 |
484 |
111 |
View Full Product Details