Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GAGE4
- Entrez Gene ID
-
2576
- Gene Name
- G antigen 4
- Gene Aliases
-
CT4.4
- Chromosome Location
-
-
- Gene Symbol
- GAGE1
- Entrez Gene ID
-
2543
- Gene Name
- G antigen 1
- Gene Aliases
-
CT4.1, GAGE-1
- Chromosome Location
-
Chr.X: 49598992 - 49608536 on Build GRCh38
- Gene Symbol
- GAGE12B
- Entrez Gene ID
-
729428
- Gene Name
- G antigen 12B
- Gene Aliases
-
GAGE-12B
- Chromosome Location
-
Chr.X: 49341183 - 49529921 on Build GRCh38
- Gene Symbol
- GAGE12C
- Entrez Gene ID
-
729422
- Gene Name
- G antigen 12C
- Gene Aliases
-
GAGE-12B
- Chromosome Location
-
Chr.X: 49532177 - 49539541 on Build GRCh38
- Gene Symbol
- GAGE12D
- Entrez Gene ID
-
100132399
- Gene Name
- G antigen 12D
- Gene Aliases
-
GAGE-12B
- Chromosome Location
-
Chr.X: 49541722 - 49549107 on Build GRCh38
- Gene Symbol
- GAGE12E
- Entrez Gene ID
-
729431
- Gene Name
- G antigen 12E
- Gene Aliases
-
GAGE-12B
- Chromosome Location
-
Chr.X: 49551289 - 49558649 on Build GRCh38
- Gene Symbol
- GAGE12F
- Entrez Gene ID
-
100008586
- Gene Name
- G antigen 12F
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 49560808 - 49568208 on Build GRCh38
- Gene Symbol
- GAGE12G
- Entrez Gene ID
-
645073
- Gene Name
- G antigen 12G
- Gene Aliases
-
AL4, CT4.7, GAGE-12G
- Chromosome Location
-
Chr.X: 49570400 - 49577757 on Build GRCh38
- Gene Symbol
- GAGE12H
- Entrez Gene ID
-
729442
- Gene Name
- G antigen 12H
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 49579949 - 49587304 on Build GRCh38
- Gene Symbol
- GAGE12I
- Entrez Gene ID
-
26748
- Gene Name
- G antigen 12I
- Gene Aliases
-
AL4, CT4.7, GAGE-7B, GAGE7B
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001477.1 |
NP_001468.1 |
2 - 3 |
142 |
111 |
- Gene Symbol
- GAGE12J
- Entrez Gene ID
-
729396
- Gene Name
- G antigen 12J
- Gene Aliases
-
GAGE11
- Chromosome Location
-
Chr.X: 49322030 - 49329387 on Build GRCh38
- Gene Symbol
- GAGE13
- Entrez Gene ID
-
645051
- Gene Name
- G antigen 13
- Gene Aliases
-
GAGE-12A, GAGE-13, GAGE12A
- Chromosome Location
-
Chr.X: 49331603 - 49338952 on Build GRCh38
- Gene Symbol
- GAGE3
- Entrez Gene ID
-
2575
- Gene Name
- G antigen 3
- Gene Aliases
-
CT4.3
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U19144.1 |
- |
- |
190 |
197 |
- Gene Symbol
- GAGE5
- Entrez Gene ID
-
2577
- Gene Name
- G antigen 5
- Gene Aliases
-
CT4.5
- Chromosome Location
-
-
- Gene Symbol
- GAGE6
- Entrez Gene ID
-
2578
- Gene Name
- G antigen 6
- Gene Aliases
-
CT4.6
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001476.1 |
NP_001467.1 |
2 - 3 |
169 |
111 |
GenBank mRNA |
U19147.1 |
- |
2 - 3 |
169 |
111 |
- Gene Symbol
- GAGE7
- Entrez Gene ID
-
2579
- Gene Name
- G antigen 7
- Gene Aliases
-
AL4, CT4.7, GAGE-7
- Chromosome Location
-
-
View Full Product Details