Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC37A
Entrez Gene ID
9884
Gene Name
leucine rich repeat containing 37A
Gene Aliases
LRRC37
Chromosome Location
Chr.17: 46248044 - 46339367 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014834.4 NP_055649.4 5 - 6 2913 78
XM_005257879.4 XP_005257936.1 5 - 6 3171 78
XM_005257882.4 XP_005257939.1 6 - 7 3357 78
XM_005257883.4 XP_005257940.1 5 - 6 3171 78
XM_017025472.1 XP_016880961.1 7 - 8 2770 78
XM_017025473.1 XP_016880962.1 5 - 6 417 78
GenBank mRNA BC040501.1 - 5 - 6 2913 78
CR749232.1 - 5 - 6 2914 78
JQ790566.1 - 5 - 6 470 78
JQ790572.1 - 5 - 6 470 78
JQ790578.1 - 5 - 6 470 78
JQ790579.1 - 5 - 6 470 78
JQ790580.1 - 5 - 6 470 78
JQ790581.1 - 5 - 6 470 78
JQ790688.1 - 5 - 6 470 78
JQ790690.1 - 5 - 6 470 78
JQ790691.1 - 5 - 6 470 78
JQ790692.1 - 5 - 6 470 78
JQ790693.1 - 5 - 6 470 78
JQ790694.1 - 5 - 6 470 78
JQ790699.1 - 5 - 6 470 78
JQ790726.1 - 5 - 6 472 78
JQ790727.1 - 5 - 6 472 78
JQ790729.1 - 5 - 6 472 78
JQ790730.1 - 5 - 6 472 78
JQ790738.1 - 5 - 6 472 78
JQ790743.1 - 5 - 6 472 78
JQ790744.1 - 5 - 6 471 78
JQ790749.1 - 5 - 6 472 78
JQ790752.1 - 5 - 6 472 78
JQ790755.1 - 5 - 6 472 78
JQ790756.1 - 5 - 6 472 78
JQ790757.1 - 5 - 6 472 78
JQ790758.1 - 5 - 6 472 78

Gene Symbol
LRRC37A2
Entrez Gene ID
474170
Gene Name
leucine rich repeat containing 37 member A2
Gene Aliases
LRRC37
Chromosome Location
Chr.17: 46512657 - 46556747 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006607.2 NP_001006608.2 5 - 6 2913 78
XM_011524841.2 XP_011523143.1 4 - 5 2894 78
XM_011524842.2 XP_011523144.1 5 - 6 2966 78
XM_011524849.1 XP_011523151.1 5 - 6 2966 78
GenBank mRNA AK126994.1 - 5 - 6 1595 78
AY386262.1 - 5 - 6 1334 78
BC144427.1 - 5 - 6 576 78
BC144428.1 - 5 - 6 576 78
JQ790563.1 - 5 - 6 470 78
JQ790564.1 - 5 - 6 470 78
JQ790568.1 - 5 - 6 470 78
JQ790570.1 - 5 - 6 470 78
JQ790571.1 - 5 - 6 470 78
JQ790576.1 - 5 - 6 470 78
JQ790598.1 - 4 - 5 398 78
JQ790617.1 - 5 - 6 470 78
JQ790626.1 - 5 - 6 470 78
JQ790700.1 - 5 - 6 470 78
JQ790703.1 - 5 - 6 446 78
JQ790739.1 - 5 - 6 472 78
JQ790740.1 - 5 - 6 472 78
JQ790750.1 - 5 - 6 472 78
JQ790764.1 - 5 - 6 472 78

Gene Symbol
LRRC37A3
Entrez Gene ID
374819
Gene Name
leucine rich repeat containing 37 member A3
Gene Aliases
LRRC37, LRRC37A
Chromosome Location
Chr.17: 64854147 - 64966228 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JQ790734.1 - 5 - 6 480 78
JQ790762.1 - 5 - 6 473 78

Gene Symbol
LRRC37A4P
Entrez Gene ID
55073
Gene Name
leucine rich repeat containing 37 member A4, pseudogene
Gene Aliases
LRRC37, LRRC37A4
Chromosome Location
Chr.17: 45505883 - 45520523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JQ790698.1 - 5 - 6 470 78

View Full Product Details