The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016334.3 | NP_057418.1 | 11 - 12 | 1152 | 103 |
XM_005277401.3 | XP_005277458.1 | 11 - 12 | 1125 | 103 | |
XM_005277402.4 | XP_005277459.1 | 11 - 12 | 1289 | 103 | |
XM_005277404.3 | XP_005277461.1 | 11 - 12 | 1145 | 103 | |
XM_006711377.3 | XP_006711440.1 | 12 - 13 | 1188 | 103 | |
XM_006711378.2 | XP_006711441.1 | 12 - 13 | 1191 | 103 | |
XM_011509610.2 | XP_011507912.1 | 12 - 13 | 1185 | 103 | |
XM_011509612.2 | XP_011507914.1 | 12 - 13 | 1146 | 103 | |
XM_017001448.1 | XP_016856937.1 | 12 - 13 | 1688 | 103 | |
GenBank mRNA | AF151035.1 | - | 3 - 4 | 264 | 103 |
AK299995.1 | - | 11 - 12 | 1123 | 103 | |
AK302266.1 | - | 11 - 12 | 1144 | 103 | |
AL133020.1 | - | 11 - 12 | 1653 | 103 | |
CD102528.1 | - | 6 - 7 | 517 | 103 | |
KJ534849.1 | - | 11 - 12 | 1014 | 103 | |
KJ534850.1 | - | 11 - 12 | 1008 | 103 | |
KJ534851.1 | - | 11 - 12 | 939 | 103 | |
U78723.1 | - | 11 - 12 | 1111 | 103 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001097612.1 | NP_001091081.1 | 11 - 12 | 1182 | 103 |
NM_001097613.2 | NP_001091082.2 | 12 - 13 | 1228 | 103 | |
NR_036541.1 | 11 - 12 | 1202 | 103 | ||
XM_006711492.3 | XP_006711555.1 | 11 - 12 | 1152 | 103 | |
XM_006711493.3 | XP_006711556.1 | 12 - 13 | 1160 | 103 | |
XM_011509908.2 | XP_011508210.1 | 12 - 13 | 1198 | 103 | |
XM_011509909.2 | XP_011508211.1 | 11 - 12 | 1410 | 103 | |
GenBank mRNA | AB097024.1 | - | 11 - 12 | 1123 | 103 |
AB097025.1 | - | 11 - 12 | 1123 | 103 | |
AF132947.1 | - | 11 - 12 | 1107 | 103 | |
AK001354.1 | - | 4 - 5 | 1920 | 103 | |
AK021758.1 | - | 11 - 12 | 1172 | 103 | |
AK094228.1 | - | 2 - 3 | 1123 | 103 | |
AK301939.1 | - | 11 - 12 | 1029 | 103 | |
AK316371.1 | - | 12 - 13 | 1140 | 103 | |
AY358966.1 | - | 11 - 12 | 1157 | 103 | |
BC003187.1 | - | 11 - 12 | 1085 | 103 | |
BC067816.1 | - | 11 - 12 | 1141 | 103 | |
BC146880.1 | - | 11 - 12 | 1103 | 103 | |
BT006679.1 | - | 11 - 12 | 1008 | 103 | |
KJ534847.1 | - | 11 - 12 | 1008 | 103 | |
KJ534848.1 | - | 11 - 12 | 1014 | 103 | |
KJ535029.1 | - | 10 - 11 | 1008 | 103 | |
KJ535049.1 | - | 11 - 12 | 1028 | 103 |