The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123391.3 | NP_001116863.3 | 12 - 13 | 1040 | 108 |
GenBank mRNA | AB449884.1 | - | 11 - 12 | 928 | 108 |
AK302064.1 | - | 13 - 14 | 1266 | 108 | |
AK303259.1 | - | 11 - 12 | 1060 | 108 | |
AX775941.1 | - | 14 - 15 | 1289 | 108 | |
AX775943.1 | - | 14 - 15 | 1289 | 108 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291465.1 | NP_001278394.1 | 12 - 13 | 1028 | 108 |
XM_006722248.2 | XP_006722311.1 | 11 - 12 | 2534 | 108 | |
XM_006722249.2 | XP_006722312.1 | 10 - 11 | 2468 | 108 | |
XM_006722252.3 | XP_006722315.1 | 10 - 11 | 2285 | 108 | |
XM_011524176.2 | XP_011522478.1 | 12 - 13 | 1141 | 108 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001417.6 | NP_001001417.6 | 12 - 13 | 1074 | 108 |
XM_005257980.4 | XP_005258037.2 | 12 - 13 | 1204 | 108 | |
GenBank mRNA | AF540953.1 | - | 11 - 12 | 928 | 108 |
AK292257.1 | - | 12 - 13 | 1045 | 108 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001418.6 | NP_001001418.5 | 12 - 13 | 1068 | 108 |
XM_006721904.2 | XP_006721967.1 | 11 - 12 | 2534 | 108 | |
XM_006721905.2 | XP_006721968.1 | 10 - 11 | 2468 | 108 | |
XM_006721907.3 | XP_006721970.1 | 10 - 11 | 2285 | 108 | |
XM_011524814.2 | XP_011523116.1 | 12 - 13 | 1141 | 108 | |
GenBank mRNA | AK313329.1 | - | 12 - 13 | 1068 | 108 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291466.1 | NP_001278395.1 | 12 - 13 | 1026 | 108 |
XM_006722254.1 | XP_006722317.1 | 15 - 16 | 1375 | 108 | |
GenBank mRNA | AL136860.1 | - | 12 - 13 | 1026 | 108 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032258.4 | NP_115634.3 | 12 - 13 | 1042 | 108 |
GenBank mRNA | AK225985.1 | - | 12 - 13 | 1033 | 108 |
AK292344.1 | - | 12 - 13 | 1085 | 108 | |
BC075809.1 | - | 12 - 13 | 1074 | 108 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291462.1 | NP_001278391.1 | 12 - 13 | 1028 | 108 |
XM_005276914.3 | XP_005276971.1 | 12 - 13 | 2853 | 108 | |
GenBank mRNA | AK292308.1 | - | 13 - 14 | 1078 | 108 |
AK301823.1 | - | 10 - 11 | 840 | 108 | |
BC071680.1 | - | 12 - 13 | 1028 | 108 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123392.3 | NP_001116864.3 | 12 - 13 | 1042 | 108 |
XM_006722055.1 | XP_006722118.1 | 11 - 12 | 1682 | 108 | |
XM_006722056.1 | XP_006722119.1 | 10 - 11 | 1616 | 108 | |
XM_011525172.1 | XP_011523474.1 | 11 - 12 | 1865 | 108 | |
XM_011525173.2 | XP_011523475.1 | 12 - 13 | 1236 | 108 | |
XM_011525174.1 | XP_011523476.1 | 10 - 11 | 1799 | 108 | |
GenBank mRNA | BM844606.1 | - | 3 - 4 | 240 | 108 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291463.1 | NP_001278392.1 | 12 - 13 | 1028 | 108 |
XM_006722223.3 | XP_006722286.1 | 12 - 13 | 1299 | 108 | |
XM_006722224.3 | XP_006722287.1 | 11 - 12 | 2544 | 108 | |
XM_006722226.3 | XP_006722289.1 | 10 - 11 | 2478 | 108 | |
XM_011524183.2 | XP_011522485.1 | 10 - 11 | 2661 | 108 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291464.1 | NP_001278393.1 | 12 - 13 | 1028 | 108 |
XM_006722235.2 | XP_006722298.1 | 11 - 12 | 3306 | 108 | |
XM_006722236.1 | XP_006722299.1 | 12 - 13 | 1878 | 108 | |
XM_006722237.2 | XP_006722300.1 | 11 - 12 | 3123 | 108 | |
XM_006722239.2 | XP_006722302.1 | 10 - 11 | 3057 | 108 | |
XM_011524172.1 | XP_011522474.1 | 10 - 11 | 3240 | 108 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291459.1 | NP_001278388.1 | 12 - 13 | 1042 | 108 |
XM_006722242.3 | XP_006722305.1 | 11 - 12 | 4282 | 108 | |
XM_006722243.2 | XP_006722306.1 | 10 - 11 | 2468 | 108 | |
XM_006722246.3 | XP_006722309.1 | 10 - 11 | 1616 | 108 | |
XM_011524175.2 | XP_011522477.1 | 12 - 13 | 1141 | 108 | |
GenBank mRNA | AK095385.1 | - | 10 - 11 | 1616 | 108 |
AK122833.1 | - | 8 - 9 | 2353 | 108 | |
AX775949.1 | - | 10 - 11 | 1616 | 108 | |
AX775951.1 | - | 10 - 11 | 1616 | 108 |