The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001418.6 | NP_001001418.5 | 4 - 5 | 335 | 92 |
XM_006721904.2 | XP_006721967.1 | 3 - 4 | 1801 | 92 | |
XM_006721905.2 | XP_006721968.1 | 3 - 4 | 1801 | 92 | |
XM_006721907.3 | XP_006721970.1 | 3 - 4 | 1618 | 92 | |
XM_006721908.2 | XP_006721971.1 | 3 - 4 | 1801 | 92 | |
XM_011524814.2 | XP_011523116.1 | 4 - 5 | 408 | 92 | |
XM_017024646.1 | XP_016880135.1 | 3 - 4 | 1801 | 92 | |
GenBank mRNA | AK313329.1 | - | 4 - 5 | 335 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291465.1 | NP_001278394.1 | 4 - 5 | 295 | 92 |
XM_006722248.2 | XP_006722311.1 | 3 - 4 | 1801 | 92 | |
XM_006722249.2 | XP_006722312.1 | 3 - 4 | 1801 | 92 | |
XM_006722250.2 | XP_006722313.1 | 3 - 4 | 1801 | 92 | |
XM_006722252.3 | XP_006722315.1 | 3 - 4 | 1618 | 92 | |
XM_006722253.2 | XP_006722316.1 | 3 - 4 | 1801 | 92 | |
XM_011524176.2 | XP_011522478.1 | 4 - 5 | 408 | 92 | |
XM_017024010.1 | XP_016879499.1 | 3 - 4 | 1801 | 92 | |
GenBank mRNA | BI088323.1 | - | 2 - 3 | 58 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123391.3 | NP_001116863.3 | 4 - 5 | 307 | 92 |
GenBank mRNA | AB449884.1 | - | 3 - 4 | 195 | 92 |
AK302064.1 | - | 5 - 6 | 533 | 92 | |
AK303259.1 | - | 3 - 4 | 327 | 92 | |
AX775941.1 | - | 6 - 7 | 556 | 92 | |
AX775943.1 | - | 6 - 7 | 556 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001417.6 | NP_001001417.6 | 4 - 5 | 341 | 92 |
XM_005257980.4 | XP_005258037.2 | 4 - 5 | 471 | 92 | |
GenBank mRNA | AF540953.1 | - | 3 - 4 | 195 | 92 |
AK292257.1 | - | 4 - 5 | 312 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291466.1 | NP_001278395.1 | 4 - 5 | 293 | 92 |
XM_006722254.1 | XP_006722317.1 | 7 - 8 | 642 | 92 | |
GenBank mRNA | AL136860.1 | - | 4 - 5 | 293 | 92 |
BC047739.1 | - | 5 - 6 | 506 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032258.4 | NP_115634.3 | 4 - 5 | 309 | 92 |
GenBank mRNA | AK225985.1 | - | 4 - 5 | 300 | 92 |
AK292344.1 | - | 4 - 5 | 352 | 92 | |
BC075809.1 | - | 4 - 5 | 341 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291462.1 | NP_001278391.1 | 4 - 5 | 295 | 92 |
XM_005276914.3 | XP_005276971.1 | 4 - 5 | 2120 | 92 | |
GenBank mRNA | AK292308.1 | - | 4 - 5 | 345 | 92 |
AK301823.1 | - | 4 - 5 | 294 | 92 | |
BC071680.1 | - | 4 - 5 | 295 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123392.3 | NP_001116864.3 | 4 - 5 | 309 | 92 |
XM_006722055.1 | XP_006722118.1 | 3 - 4 | 949 | 92 | |
XM_006722056.1 | XP_006722119.1 | 3 - 4 | 949 | 92 | |
XM_011525172.1 | XP_011523474.1 | 3 - 4 | 1132 | 92 | |
XM_011525173.2 | XP_011523475.1 | 4 - 5 | 503 | 92 | |
XM_011525174.1 | XP_011523476.1 | 3 - 4 | 1132 | 92 | |
XM_011525175.1 | XP_011523477.1 | 3 - 4 | 1132 | 92 | |
XM_011525176.1 | XP_011523478.1 | 3 - 4 | 1132 | 92 | |
GenBank mRNA | CB131287.1 | - | 3 - 4 | 145 | 92 |
DC398423.1 | - | 4 - 5 | 310 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291463.1 | NP_001278392.1 | 4 - 5 | 295 | 92 |
XM_006722223.3 | XP_006722286.1 | 4 - 5 | 566 | 92 | |
XM_006722224.3 | XP_006722287.1 | 3 - 4 | 1811 | 92 | |
XM_006722226.3 | XP_006722289.1 | 3 - 4 | 1811 | 92 | |
XM_011524183.2 | XP_011522485.1 | 3 - 4 | 1994 | 92 | |
XM_011524184.2 | XP_011522486.1 | 3 - 4 | 1994 | 92 | |
XM_011524185.2 | XP_011522487.1 | 3 - 4 | 1994 | 92 | |
GenBank mRNA | BC078140.1 | - | 4 - 5 | 296 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291464.1 | NP_001278393.1 | 4 - 5 | 295 | 92 |
XM_006722235.2 | XP_006722298.1 | 3 - 4 | 2573 | 92 | |
XM_006722236.1 | XP_006722299.1 | 4 - 5 | 1145 | 92 | |
XM_006722237.2 | XP_006722300.1 | 3 - 4 | 2390 | 92 | |
XM_006722239.2 | XP_006722302.1 | 3 - 4 | 2390 | 92 | |
XM_011524172.1 | XP_011522474.1 | 3 - 4 | 2573 | 92 | |
XM_011524173.1 | XP_011522475.1 | 3 - 4 | 2573 | 92 | |
XM_011524174.1 | XP_011522476.1 | 3 - 4 | 2573 | 92 | |
GenBank mRNA | DC397859.1 | - | 4 - 5 | 328 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291459.1 | NP_001278388.1 | 4 - 5 | 309 | 92 |
XM_006722242.3 | XP_006722305.1 | 3 - 4 | 3549 | 92 | |
XM_006722243.2 | XP_006722306.1 | 3 - 4 | 1801 | 92 | |
XM_006722244.2 | XP_006722307.1 | 3 - 4 | 1801 | 92 | |
XM_006722246.3 | XP_006722309.1 | 3 - 4 | 949 | 92 | |
XM_006722247.2 | XP_006722310.1 | 3 - 4 | 1801 | 92 | |
XM_011524175.2 | XP_011522477.1 | 4 - 5 | 408 | 92 | |
GenBank mRNA | AK095385.1 | - | 3 - 4 | 949 | 92 |
AX775949.1 | - | 3 - 4 | 949 | 92 | |
AX775951.1 | - | 3 - 4 | 949 | 92 |