Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TBC1D3B
- Entrez Gene ID
-
414059
- Gene Name
- TBC1 domain family member 3B
- Gene Aliases
-
PRC17, TBC1D3I, TBC1D3L
- Chromosome Location
-
Chr.17: 36165681 - 36176624 on Build GRCh38
- Gene Symbol
- TBC1D3G
- Entrez Gene ID
-
101060321
- Gene Name
- TBC1 domain family member 3G
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 36322242 - 36334759 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK292308.1 |
- |
11 - 12 |
913 |
113 |
View Full Product Details