Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRAMEF15
Entrez Gene ID
653619
Gene Name
PRAME family member 15
Gene Aliases
-
Chromosome Location
Chr.1: 13315581 - 13322595 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098376.2 NP_001091846.1 2 - 3 381 79

Gene Symbol
PRAMEF25
Entrez Gene ID
441873
Gene Name
PRAME family member 25
Gene Aliases
-
Chromosome Location
Chr.1: 13073678 - 13078038 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001310134.1 NP_001297063.1 1 - 2 79

Gene Symbol
PRAMEF26
Entrez Gene ID
645359
Gene Name
PRAME family member 26
Gene Aliases
-
Chromosome Location
Chr.1: 13148905 - 13153111 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306072.1 NP_001293001.1 1 - 2 79

Gene Symbol
PRAMEF9
Entrez Gene ID
343070
Gene Name
PRAME family member 9
Gene Aliases
-
Chromosome Location
Chr.1: 13175281 - 13179132 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK124292.1 - 2 - 3 381 79
BC101343.2 - 2 - 3 367 79
BC137391.1 - 2 - 3 367 79
BC171794.1 - 2 - 3 367 79

View Full Product Details