Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SUPT20HL1
- Entrez Gene ID
-
100130302
- Gene Name
- SPT20 homolog, SAGA complex component-like 1
- Gene Aliases
-
FAM48B1, SPT20L
- Chromosome Location
-
Chr.X: 24362761 - 24365424 on Build GRCh38
- Gene Symbol
- SUPT20HL2
- Entrez Gene ID
-
170067
- Gene Name
- SPT20 homolog, SAGA complex component-like 2
- Gene Aliases
-
FAM48B1, FAM48B2
- Chromosome Location
-
Chr.X: 24310862 - 24313315 on Build GRCh38
View Full Product Details