Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXO3
Entrez Gene ID
2309
Gene Name
forkhead box O3
Gene Aliases
AF6q21, FKHRL1, FKHRL1P2, FOXO2, FOXO3A
Chromosome Location
Chr.6: 108559823 - 108684769 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001455.3 NP_001446.1 1 - 1 56 90
GenBank mRNA AF032886.1 - 1 - 1 637 90
AK092357.1 - 1 - 1 42 90
BC068552.1 - 1 - 1 56 90

View Full Product Details