Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
MAP2K2
Entrez Gene ID
5605
Gene Name
mitogen-activated protein kinase kinase 2
Gene Aliases
CFC4, MAPKK2, MEK2, MKK2, PRKMK2
Chromosome Location
Chr.19: 4090321 - 4124184 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030662.3 NP_109587.1 7 - 7 1155 74
XM_017026989.1 XP_016882478.1 7 - 7 1210 74
GenBank mRNA AK093125.1 - 4 - 4 1202 74
AK126446.1 - 1 - 1 1460 74
BC000471.2 - 7 - 7 1155 74
BC018645.2 - 7 - 7 1097 74
BX538081.1 - 8 - 8 905 74
L11285.1 - 7 - 7 985 74

View Full Product Details