The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145040.2 | NP_659477.2 | 2 - 2 | 952 | 102 |
GenBank mRNA | AF339881.1 | - | 2 - 2 | 954 | 102 |
BC011585.1 | - | 2 - 2 | 952 | 102 |