The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002700.2 | NP_002691.1 | 1 - 1 | 50 | 93 |
GenBank mRNA | BC104923.1 | - | 1 - 1 | 50 | 93 |
BC112207.1 | - | 1 - 1 | 50 | 93 |