The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002995.2 | NP_002986.1 | 3 - 3 | 998 | 176 |
GenBank mRNA | D43768.1 | - | 3 - 3 | 853 | 176 |